Anti-IgG Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
619-106-002
RL619-106-002EA 209.8 USD
RL619-106-002
Anti-IgG Goat Polyclonal Antibody (Biotin)
Antibodies
Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Armenian Hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR