Anti-TCR g/d Armenian Hamster Monoclonal Antibody (APC (Allophycocyanin)) [clone: GL3]

Supplier: BioLegend
118116 118115
118116-BLEA 224.93 USD
118116-BL 118115-BL
Anti-TCR g/d Armenian Hamster Monoclonal Antibody (APC (Allophycocyanin)) [clone: GL3]
Antibodies
APC anti-mouse TCR γ/δ [GL3]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: TCR g/d
Clonality: Monoclonal
Clone: GL3
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR