Anti-IL-6 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: MP5-20F3]

Supplier: BioLegend
504508 504507
504507-BLEA 110.1 USD
504507-BL 504508-BL
Anti-IL-6 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: MP5-20F3]
Antibodies
APC anti-mouse IL-6 [MP5-20F3]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ICFC; Size: 100 μg

The MP5-20F3 antibody reacts with mouse interleukin-6 (IL-6). The MP5-20F3 antibody can neutralize the bioactivity of natural or recombinant IL-6. Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-6
Clonality: Monoclonal
Clone: MP5-20F3
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR