Anti-IgM, k Isotype Ctrl Rat Monoclonal Antibody (Alexa Fluor® 647) [clone: RTK2118]

Supplier: BioLegend
400813
400813-BLEA 109.03 USD
400813-BL
Anti-IgM, k Isotype Ctrl Rat Monoclonal Antibody (Alexa Fluor® 647) [clone: RTK2118]
Antibodies
Alexa Fluor® 647 Rat IgM, κ Isotype Ctrl [RTK2118]; Isotype: Rat IgM, κ; Reactivity: Rat; Apps: FC; Size: 25 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis as negative control, and the purity is greater than 95% by SDS-PAGE. Use at concentrations comparable to those of the specific antibody of interest.

Type: Control
Antigen: IgM, k Isotype Ctrl
Clonality: Monoclonal
Clone: RTK2118
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rat
Isotype: Rat IgM, κ
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR