Anti-RAE-1gamma Rat Monoclonal Antibody (Alexa Fluor® 647) [clone: CX1]

Supplier: BioLegend
130110
130110-BLEA 266.77 USD
130110-BL
Anti-RAE-1gamma Rat Monoclonal Antibody (Alexa Fluor® 647) [clone: CX1]
Antibodies
Alexa Fluor® 647 anti-mouse RAE-1γ [CX1]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: RAE-1gamma
Clonality: Monoclonal
Clone: CX1
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR