Anti-AKT1 Mouse Monoclonal Antibody [clone: 17F6.B11]

Supplier: Rockland Immunochemicals
200-301-B19
RL200-301-B19EA 2703.78 USD
RL200-301-B19
Anti-AKT1 Mouse Monoclonal Antibody [clone: 17F6.B11]
Antibodies
This monoclonal antibody is suitable for ELISA, immunochemistry, immunoprecipitation, immunofluorescent microscopy, and western blotting.  Expect a band approximately 56 kDa in size corresponding to phosphorylated AKT Protein by western blotting.

Recommended Dilutions: ELISA: 1:20000; Western Blot: 1:500 - 1:3000; Immunochemistry: 20 µg/ml;

Type: Primary
Antigen: AKT1
Clonality: Monoclonal
Clone: 17F6.B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR