Anti-AKR1C2 Rabbit Polyclonal Antibody

A01054-40UG
95044-220EA 119.82 USD
95044-220
Anti-AKR1C2 Rabbit Polyclonal Antibody
Type: Primary
Antigen: AKR1C2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR