Anti-5HT5A Rabbit Polyclonal Antibody

A01209-40UG
95045-232EA 119.82 USD
95045-232
Anti-5HT5A Rabbit Polyclonal Antibody
Type: Primary
Antigen: 5HT5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR