Anti-5HT3 Rabbit Polyclonal Antibody

A01210-40UG
95045-236EA 119.82 USD
95045-236
Anti-5HT3 Rabbit Polyclonal Antibody
Type: Primary
Antigen: 5HT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR