Anti-YWHAB Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
612-401-D01
RL612-401-D01EA 518.29 USD
RL612-401-D01
Anti-YWHAB Rabbit Polyclonal Antibody
Antibodies
14-3-3 antibody is suitable for use in Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 29 kDa in size corresponding to 14-3-3 Protein in the appropriate cell lysate or extract.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: YWHAB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR