Anti-THE™ HA Tag Mouse Monoclonal Antibody

A01244-100UG
95045-688EA 359.42 USD
95045-688
Anti-THE™ HA Tag Mouse Monoclonal Antibody
Type: Primary
Antigen: HA-Tag
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR