JACKSON SAFETY® NOMEX™ Hood Winterliner, KIMBERLY-CLARK PROFESSIONAL®
14504
33001-156EA 42.6 USD
33001-156
JACKSON SAFETY® NOMEX™ Hood Winterliner, KIMBERLY-CLARK PROFESSIONAL®
Protects head and neck against flame and electrical arcs in temperatures from 45° to 32° F (8° to 0°C)
  • Protects head and neck against flame and electrical arcs in temperatures from 45° to 32° F (8° to 0°C)
  • Constructed of doublelayered 100% Nomex
  • Shoulder length, extra-wide collar can be tucked inside a jacket
  • Can be worn alone or underneath hard hat or welding helmet
  • Retains FR protection throughout life of the product
Constructed of doublelayered 100% Nomex. Shoulder length, extra wide collar can be tucked inside a jacket. Can be worn alone or underneath a hard hat or welding helmet. Retains FR protection throughout the life of the product.