SCIENCEWARE® Blow-Hard® O.S. Extra™ Dust Remover, Bel-Art
170800200
47750-915EA 47.03 USD
47750-915
SCIENCEWARE® Blow-Hard® O.S. Extra™ Dust Remover, Bel-Art
Provides powerful blasts of triple filtered air for cleaning a wide variety of laboratory equipment which could be damaged by wiping, such as microscopes.
  • Delivers triple-filtered air
  • Handy extension tube
  • Ozone-safe
  • Non-flammable
A handy extension tube (included) effectively penetrates hard-to-reach places for thorough dust removal.