Anti-HP Goat Polyclonal Antibody

Supplier: GeneTex
GTX77180
89353-528EA 648.3 USD
89353-528
Anti-HP Goat Polyclonal Antibody
Antibodies
Affinity Purified Goat anti - Bovine Haptoglobin

WB: 1:1000-1:5000; ELISA: coating 1:200-1:500. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher

Type: Primary
Antigen: HP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR