Anti-STAU1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX106566
89354-734EA 690.66 USD
89354-734
Anti-STAU1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Staufen (staufen, RNA binding protein, homolog 1 (Drosophila))

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IP: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: STAU1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR