Anti-RAD51AP1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX106411
89354-674EA 582.76 USD
89354-674
Anti-RAD51AP1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to RAD51AP1 (RAD51 associated protein 1)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: RAD51AP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR