Anti-RAB2A Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX106337
89354-630EA 448.72 USD
89354-630
Anti-RAB2A Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to RAB2A (RAB2A, member RAS oncogene family)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: RAB2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR