Anti-TOM1L2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX106295
89354-586EA 582.76 USD
89354-586
Anti-TOM1L2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to TOM1-like protein 2 (target of myb1-like 2 (chicken))

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: TOM1L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR