Anti-CREB Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX10564
89354-318EA 611.89 USD
89354-318
Anti-CREB Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to CREB ( phospho S129 + S133 )

WB: Use at a dilution of 1/1000. Detects a band of approximately 43 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CREB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR