Anti-CCND1 Mouse Monoclonal Antibody [clone: DCS-6]

Supplier: GeneTex
GTX10540
89354-234EA 562.37 USD
89354-234
Anti-CCND1 Mouse Monoclonal Antibody [clone: DCS-6]
Antibodies
Mouse monoclonal [DCS - 6] to Cyclin D1

  • ICC: Use at an assay dependent dilution. IHC-P: Use at an assay dependent dilution. Perform heat mediated antigen retrieval via the microwave method before commencing with IHC staining protocol. IP: Use at an assay dependent dilution. WB: Use at a dilution of 1/200. Detects a band of approximately 36 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CCND1
Clonality: Monoclonal
Clone: DCS-6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR