Anti-IRAK1BP1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX104392
89350-316EA 582.76 USD
89350-316
Anti-IRAK1BP1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to IRAK1BP1 (interleukin-1 receptor-associated kinase 1 binding protein 1)

Suggested starting dilutions are as follows: IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: IRAK1BP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR