Anti-RELA Mouse Monoclonal Antibody [clone: NF-12]

Supplier: GeneTex
GTX10437
89350-306EA 625.75 USD
89350-306
Anti-RELA Mouse Monoclonal Antibody [clone: NF-12]
Antibodies
Mouse monoclonal antibody [NF-12] to NFkB p65

For WB: Use at a dilution of 1:1,000. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: RELA
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR