Anti-TOMM22 Mouse Monoclonal Antibody [clone: 1C9-2]

Supplier: GeneTex
GTX10436
89350-300EA 638.16 USD
89350-300
Anti-TOMM22 Mouse Monoclonal Antibody [clone: 1C9-2]
Antibodies
Mouse monoclonal [1C9 - 2] to TOMM22

ICC: Use at an assay dependent dilution. WB: Use at a concentration of 0.5 - 1.0 ?g/ml. Detects a band of approximately 22 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TOMM22
Clonality: Monoclonal
Clone: 1C9-2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR