Anti-BNIP3 Mouse Monoclonal Antibody [clone: ANa40]

Supplier: GeneTex
GTX10433
89350-292EA 613.75 USD
89350-292
Anti-BNIP3 Mouse Monoclonal Antibody [clone: ANa40]
Antibodies
Mouse monoclonal [ANa40] to BNIP3

ELISA: Use at an assay dependent dilution. ICC: Use at an assay dependent dilution. IP: Use at an assay dependent dilution. WB: Use at a concentration of 2 - 4 ?g/ml. Detects a band of approximately 30 and 60 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: BNIP3
Clonality: Monoclonal
Clone: ANa40
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR