Anti-FRS2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX10425
89350-270EA 616.7 USD
89350-270
Anti-FRS2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to FRS2

For IP: Use 10?g to 20ug using 3 mg RIPA lysate of mouse NIH/3T3 cells. For WB: Use at a dilution of 1/1000. Detects a band of approximately 92 and 95 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: FRS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR