Anti-FKBP8 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX104240
89350-268EA 1199.11 USD
89350-268
Anti-FKBP8 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to FKBP8 (FK506 binding protein 8, 38kDa)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: FKBP8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR