Anti-FGF2 Mouse Monoclonal Antibody [clone: FB-8]

Supplier: GeneTex
GTX10419
89350-262EA 611.61 USD
89350-262
Anti-FGF2 Mouse Monoclonal Antibody [clone: FB-8]
Antibodies
Mouse monoclonal [FB - 8] to FGF2

Dot: Use at an assay dependent dilution. ELISA: Use at an assay dependent dilution. Neut: Use at an assay dependent dilution. RIA: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Detects a band of approximately 18 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: FGF2
Clonality: Monoclonal
Clone: FB-8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR