Anti-EGFR Mouse Monoclonal Antibody [clone: F4]

Supplier: GeneTex
GTX10415
89350-256EA 526.99 USD
89350-256
Anti-EGFR Mouse Monoclonal Antibody [clone: F4]
Antibodies
Mouse monoclonal [F4] to EGFR

ELISA: Use at an assay dependent dilution. IHC-P: Use at an assay dependent dilution. IP: Use at an assay dependent dilution. WB: Use at a dilution of 1/2000. Detects a band of approximately 170 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone: F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR