Anti-CLCN5 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX104090
89350-232EA 468.31 USD
89350-232
Anti-CLCN5 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to chloride channel 5 (chloride channel 5 (nephrolithiasis 2, X-linked, Dent disease))

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CLCN5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR