Anti-EGF Mouse Monoclonal Antibody [clone: EGF-10]

Supplier: GeneTex
GTX10409
89350-230EA 688.64 USD
89350-230
Anti-EGF Mouse Monoclonal Antibody [clone: EGF-10]
Antibodies
Mouse monoclonal [EGF - 10] to EGF

Dot: Use at a concentration of 1 ?g/ml. ELISA: Use at an assay dependent dilution. IHC-P: Use at an assay dependent dilution. RIA: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 144 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: EGF
Clonality: Monoclonal
Clone: EGF-10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR