Anti-UCHL1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX10404
89350-206EA 628.47 USD
89350-206
Anti-UCHL1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to PGP9.5 - Neuronal Marker

For IHC-Fr: Use at a dilution of 1:2,000. For WB: Use at a dilution of 1:1,000. Predicted molecular weight: 26.8 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: UCHL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR