Anti-CCR5 Mouse Monoclonal Antibody [clone: 45523.111]

Supplier: GeneTex
GTX10397
89350-178EA 482.95 USD
89350-178
Anti-CCR5 Mouse Monoclonal Antibody [clone: 45523.111]
Antibodies
Mouse monoclonal [45523.111] to CCR5

FACS: Use at a concentration of 5 - 10 ?g/ml. Determined using 10 ?l human CCR5 transfected cells (1-2.5 x 10^5 cells) in a toatal reaction volume not exceeding 200 ?l. IHC: Use at a concentration of 5 ?g/ml for 2% formaldehyde fixed cells. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CCR5
Clonality: Monoclonal
Clone: 45523.111
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR