Anti-CNP Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX103954
89350-168EA 582.56 USD
89350-168
Anti-CNP Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to CNP (2',3'-cyclic nucleotide 3' phosphodiesterase)

Suggested starting dilutions are as follows: WB: 1:1000-1:10000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CNP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR