Anti-CD55 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX103951
89350-166EA 582.76 USD
89350-166
Anti-CD55 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to CD55 (CD55 molecule, decay accelerating factor for complement (Cromer blood group))

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CD55
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR