Anti-S100 Mouse Monoclonal Antibody [clone: 6G1]

Supplier: GeneTex
GTX10203
89350-688EA 592.61 USD
89350-688
Anti-S100 Mouse Monoclonal Antibody [clone: 6G1]
Antibodies
Mouse monoclonal [6G1] to S100

ELISA: Use at an assay dependent dilution. IA: Use at an assay dependent dilution. P: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 0 kDa. Can be paired for ELISA with GTX28330 Mouse monoclonal [8B10] to S100 - Astrocyte Marker and GTX28334 Mouse monoclonal [3B10] to S100 - Astrocyte Marker . Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: S100
Clonality: Monoclonal
Clone: 6G1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR