Anti-SPHKAP Rabbit Polyclonal Antibody

GTX101951
89349-376EA 176.03 USD
89349-376
Anti-SPHKAP Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to SPHKAP (SPHK1 interactor, AKAP domain containing)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: SPHKAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR