Anti-PSA Mouse Monoclonal Antibody [clone: 5A6]

Supplier: GeneTex
GTX10185
89350-672EA 696.18 USD
89350-672
Anti-PSA Mouse Monoclonal Antibody [clone: 5A6]
Antibodies
Mouse monoclonal [5A6] to PSA

ELISA: Use at an assay dependent dilution. For the detection of PSA antigen use in ELISA as an antibody pair: GTX10185 coating, GTX10184 conjugate, or GTX10186 coating, GTX10185 conjugateFor the detection of free PSA use in ELISA as an antibody pair GTX10185 coating, GTX10187 conjugate GTX10187 coating, GTX10185 conjugate. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PSA
Clonality: Monoclonal
Clone: 5A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR