Anti-CYB5R3 Rabbit Polyclonal Antibody

GTX101788
89349-264EA 176.03 USD
89349-264
Anti-CYB5R3 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to CYB5R3 (cytochrome b5 reductase 3)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: CYB5R3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR