Anti-H3F3A Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX101754
89349-232EA 597.31 USD
89349-232
Anti-H3F3A Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Histone H3.3 (H3 histone, family 3A)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: H3F3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR