Anti-CNTN1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX101735
89349-220EA 582.76 USD
89349-220
Anti-CNTN1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Contactin 1 (contactin 1)

Suggested starting dilutions are as follows: IHC-Fr: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CNTN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR