Anti-GAS1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX101732
89349-216EA 582.76 USD
89349-216
Anti-GAS1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to GAS1 (growth arrest-specific 1)

Suggested starting dilutions are as follows: WB: Use at an assay dependent dilution. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: GAS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR