Anti-CYC1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX101717
89349-198EA 582.76 USD
89349-198
Anti-CYC1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Cytochrome C1 (cytochrome c-1)

Suggested starting dilutions are as follows: WB: 1:1000-1:10000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CYC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR