Anti-ENO2 Mouse Monoclonal Antibody [clone: 5A4]

Supplier: GeneTex
GTX10169
89350-656EA 355.95 USD
89350-656
Anti-ENO2 Mouse Monoclonal Antibody [clone: 5A4]
Antibodies
Mouse monoclonal [5A4] to NSE - Neuronal Marker

ELISA: Use at an assay dependent dilution. IHC-P: Use at a dilution of 1/100 - 1/300. Antigen retrieval is not essential but may optimise staining. WB: Use at an assay dependent dilution. Detects a band of approximately 45 kDa. Can be used in an ELISA pair with GTX28328 (conjugate). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: ENO2
Clonality: Monoclonal
Clone: 5A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR