Anti-AGPS Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX101396
89349-012EA 582.76 USD
89349-012
Anti-AGPS Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Alkyl-DHAP synthase (alkylglycerone phosphate synthase)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: AGPS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR