Anti-KDR Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX73967
89353-302EA 1125.41 USD
89353-302
Anti-KDR Rabbit Polyclonal Antibody
Antibodies
Polyclonal antibody to Flk-1 / KDR / VEGFR-2

This antibody may be diluted to a titer of 1:50-1:100 in an ABC method. However, depending on the staining conditions employed, we suggest that final dilution should be determined by the user. We suggest an incubation period of 30 minutes at room temperature. However, depending upon the fixation conditions and the staining system employed, optimal incubation conditions should be determined by the user. Formalin fixed paraffin embedded tissue sections require high temperature antigen unmasking with 10 mM citrate buffer, pH 6.0 prior to immunostaining. Can also be used for: Western Blotting

Type: Primary
Antigen: KDR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR