Anti-CK20 Mouse Monoclonal Antibody [clone: KS 20.8]

Supplier: GeneTex
GTX73825
89353-288EA 759.39 USD
89353-288
Anti-CK20 Mouse Monoclonal Antibody [clone: KS 20.8]
Antibodies
Monoclonal Mouse Antibody to Human Cytokeratin 20

This antibody may be diluted to a titer of 1:25-1:50 in an ABC method. However, depending on the staining conditions employed, we suggest that final dilution should be determined by the user. We suggest an incubation period of 30 minutes at room temperature. However, depending upon the fixation conditions and the staining system employed, optimal incubation should be determined by the user. Proteolytic digestion of formalin fixed paraffin tissue sections is required prior to immunostaining. Can also be used for: Western Blotting (1:50 to 1:100)

Type: Primary
Antigen: CK20
Clonality: Monoclonal
Clone: KS 20.8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR