Anti-GLI3 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX26050
89361-812EA 639.29 USD
89361-812
Anti-GLI3 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Gli3

For ELISA: Use at a dilution of 1:6,000 - 1:30,000. For IHC-Fr: Use at a concentration of 0.5 ?g/ml - 5 ?g/ml. For WB: Use at a dilution of 1:500 - 1:2,000. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: GLI3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Chicken, Dog, Monkey, Zebra Fish, Chimpanzee, Quail, Xenopus laevis
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR