Anti-SNX5 Goat Polyclonal Antibody

Supplier: GeneTex
GTX25983
89361-796EA 607.87 USD
89361-796
Anti-SNX5 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to SNX5

Approx. 48kDa band observed in lysates of Human Kidney and cell lines Jurkat and A549 lysates (calculated MW of 46.8kDa according to NP_055241.1). Recommended concentration: 1-3µg/ml. In paraffin embedded Human Breast shows cytoplasm staining of ductal epithelial cells. Cytoplasm staining is also observed in PCT of Human Kidney and in trophoblasts of Human Placenta . Recommended concentration, 3-6µg/ml. Both variants represent identical protein.

Type: Primary
Antigen: SNX5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR