Anti-IgG Goat Polyclonal Antibody

Supplier: GeneTex
GTX25745
89361-720EA 375.29 USD
89361-720
Anti-IgG Goat Polyclonal Antibody
Antibodies
Immunoglobulin G (IgG), is one of the most abundant proteins in serum with normal levels between 8-17 mg/ml in adult blood. IgG is important for our defence against microorganisms and the molecules are produced by B lymphocytes as a part of our adaptive immune response. The IgG molecule has two separate functions; to bind to the pathogen that elicited the response and to recruit other cells and molecules to destroy the antigen. The variability of the IgG pool is generated by somatic recombination and the number of specificities in an individual at a given time point is estimated to be 1011 variants.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR