Anti-C-RAF Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX24768
89361-490EA 610.65 USD
89361-490
Anti-C-RAF Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Raf1 (phospho Ser338/Tyr340)

WB: Use at a concentration of 0.5 - 1.5 ?g/ml. Predicted molecular weight: 74 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: C-RAF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR